Anti-HOCl-Oxidized Low Density Lipoprotein Rabbit Polyclonal Antibody

Lieferant: Oncogene
428035-100EA 1040 CHF
428035-100
Anti-HOCl-Oxidized Low Density Lipoprotein Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: HOCl-Oxidized Low Density Lipoprotein
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Pan
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR