Anti-Insulin Receptor (pTyr 1162/1163 ) Rabbit Polyclonal Antibody

Lieferant: Oncogene
407707-10EA 1430 CHF
407707-10
Anti-Insulin Receptor (pTyr 1162/1163 ) Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Insulin Receptor (pTyr 1162/1163 )
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR