Anti-Glutathione S-Transferase A1-1 Rabbit Polyclonal Antibody
Lieferant: Oncogene
354206-100EA 1320 CHF
354206-100
Anti-Glutathione S-Transferase A1-1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Glutathione S-Transferase A1-1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now