Anti-Dopamine D 4 Receptor Rabbit Polyclonal Antibody

Lieferant: Oncogene
324405-100EA 1110 CHF
324405-100
Anti-Dopamine D 4 Receptor Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Dopamine D 4 Receptor
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR