Anti-APOA1 Rabbit Polyclonal Antibody

Lieferant: Oncogene
178422-1EA 297 CHF
178422-1
Anti-APOA1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: APOA1 (apolipoprotein A-I)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR