Anti-SUMO-tag Mouse Monoclonal Antibody [clone: 4G11E9]

Lieferant: GenScript Biotech
GENSA01693-40EA 197 CHF
GENSA01693-40
Anti-SUMO-tag Mouse Monoclonal Antibody [clone: 4G11E9]
Antikörper
Type: Tag
Antigen: SUMO-tag
Clonality: monoclonal
Clone: 4G11E9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR