Anti-MAPT Rabbit Polyclonal Antibody
Lieferant: GenScript Biotech
GENSA00896-40EA 219 CHF
GENSA00896-40
Anti-MAPT Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MAPT (microtubule-associated protein tau)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity:
Order Now