Anti-KLRC1 Mouse Monoclonal Antibody [clone: 6A8.6F10.1A6]

Lieferant: Novus Biologicals
NOVUNBP2-26649EA 653 CHF
NOVUNBP2-26649
Anti-KLRC1 Mouse Monoclonal Antibody [clone: 6A8.6F10.1A6]
Antikörper
Type: Primary
Antigen: KLRC1 (killer cell lectin-like receptor subfamily C, member 1)
Clonality: monoclonal
Clone: 6A8.6F10.1A6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR