Anti-TLR9 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 26C593.2]
Lieferant: Novus Biologicals
NOVUNBP2-24907EA 856 CHF
NOVUNBP2-24907
Anti-TLR9 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 26C593.2]
Antikörper
Type: Primary
Antigen: TLR9 (toll-like receptor 9)
Clonality: monoclonal
Clone: 26C593.2
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Order Now