Anti-GATA4 Rabbit Polyclonal Antibody

Lieferant: Novus Biologicals
NOVUNBP2-24585EA 502 CHF
NOVUNBP2-24585
Anti-GATA4 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GATA4 (GATA binding protein 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR