Anti-DDX4 Rabbit Polyclonal Antibody

Lieferant: Novus Biologicals
NOVUNBP2-24558EA 495 CHF
NOVUNBP2-24558
Anti-DDX4 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: DDX4 (DEAD (Asp-Glu-Ala-Asp) box polypeptide 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR