Anti-NHLRC1 Rabbit Polyclonal Antibody

Lieferant: Novus Biologicals
NOVUNBP2-24474EA 495 CHF
NOVUNBP2-24474
Anti-NHLRC1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NHLRC1 (NHL repeat containing E3 ubiquitin protein ligase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Bovine, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR