Anti-VLDLR Mouse Monoclonal Antibody (DyLight® 350) [clone: 6A6]

Lieferant: Novus Biologicals
NOVUNBP1-78162UVEA 763 CHF
NOVUNBP1-78162UV
Anti-VLDLR Mouse Monoclonal Antibody (DyLight® 350) [clone: 6A6]
Antikörper
Type: Primary
Antigen: VLDLR (very low density lipoprotein receptor)
Clonality: monoclonal
Clone: 6A6
Conjugation: DyLight® 350
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR