Anti-LDL Receptor Mouse Monoclonal Antibody [clone: C7]

Lieferant: Novus Biologicals
NBP1-78159 NBP1-78159SS
NOVUNBP1-78159EA 725 CHF
NOVUNBP1-78159 NOVUNBP1-78159SS
Anti-LDL Receptor Mouse Monoclonal Antibody [clone: C7]
Antikörper
Mouse Monoclonal LDL Receptor Antibody (C7). Tested Applications: Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunoprecipitation. Tested Reactivity: Human, Bovine.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:200

Type: Primary
Antigen: LDLR
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR