Anti-Lipase A Rabbit Polyclonal Antibody
Lieferant: Novus Biologicals
NBP1-54155 NBP1-54155SS
NOVUNBP1-54155EA 742 CHF
NOVUNBP1-54155 NOVUNBP1-54155SS
Anti-Lipase A Rabbit Polyclonal Antibody
Antikörper
Rabbit Polyclonal Lipase A Antibody. Tested Applications: Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human, Mouse.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:100
Type: Primary
Antigen: LIPA
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human