Anti-GAP43 Sheep Polyclonal Antibody
Lieferant: Novus Biologicals
NOVUNBP1-41123SSEA 276 CHF
NOVUNBP1-41123SS NOVUNBP1-41123
Anti-GAP43 Sheep Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GAP43 (growth associated protein 43)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: none
Reactivity:
Order Now