Anti-ACTB Rabbit Polyclonal Antibody

Lieferant: Novus Biologicals
NOVUNB600-503EA 748 CHF
NOVUNB600-503 NOVUNB600-503SS
Anti-ACTB Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ACTB (actin, beta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR