Anti-ZIC1 Rabbit Polyclonal Antibody
Lieferant: Novus Biologicals
NB600-488 NB600-488SS
NOVUNB600-488EA 716 CHF
NOVUNB600-488 NOVUNB600-488SS
Anti-ZIC1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ZIC1 (Zic family member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse