Anti-PARK7 Rabbit Polyclonal Antibody

Lieferant: Novus Biologicals
NOVUNB300-270EA 647 CHF
NOVUNB300-270 NOVUNB300-270SS
Anti-PARK7 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PARK7 (parkinson protein 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Chicken, Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR