Anti-TRPM2 Rabbit Polyclonal Antibody
Lieferant: Novus Biologicals
NB110-82364 NB110-82364SS
NOVUNB110-82364EA 645 CHF
NOVUNB110-82364 NOVUNB110-82364SS
Anti-TRPM2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TRPM2 (transient receptor potential cation channel, subfamily M, member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse, Rat