Anti-HSPA13 Mouse Monoclonal Antibody ATTO 488 [clone: C92F3A-5]

Lieferant: Novus Biologicals
NOVUNB110-61582EA 435 CHF
NOVUNB110-61582
Anti-HSPA13 Mouse Monoclonal Antibody ATTO 488 [clone: C92F3A-5]
Antikörper
Type: Primary
Antigen: HSPA13 (heat shock protein 70kDa family, member 13)
Clonality: monoclonal
Clone: C92F3A-5
Conjugation: ATTO 488
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR