Anti-ALOX5 Rabbit Polyclonal Antibody
Lieferant: Novus Biologicals
NOVUNB110-58749SSEA 292 CHF
NOVUNB110-58749SS NOVUNB110-58749
Anti-ALOX5 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ALOX5 (arachidonate 5-lipoxygenase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now