Anti-6X His tag Mouse Monoclonal Antibody [clone: AD1.1.10]

Lieferant: Novus Biologicals
NB100-64768 NB100-64768SS
NOVUNB100-64768EA 749 CHF
NOVUNB100-64768 NOVUNB100-64768SS
Anti-6X His tag Mouse Monoclonal Antibody [clone: AD1.1.10]
Antikörper
Type: Tag
Antigen: 6X His tag (6X His tag)
Clonality: monoclonal
Clone: AD1.1.10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Pan
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR