Anti-Avian Flu Nucleoprotein Rabbit Polyclonal Antibody

Lieferant: Novus Biologicals
NOVUNB100-56572EA 682 CHF
NOVUNB100-56572
Anti-Avian Flu Nucleoprotein Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Avian Flu Nucleoprotein
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR