Anti-S100A7 Mouse Monoclonal Antibody [clone: 47C1068]

Lieferant: Novus Biologicals
NOVUNB100-56559EA 716 CHF
NOVUNB100-56559
Anti-S100A7 Mouse Monoclonal Antibody [clone: 47C1068]
Antikörper
Type: Primary
Antigen: S100A7 (S100 calcium binding protein A7)
Clonality: monoclonal
Clone: 47C1068
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR