Anti-MYOD1 Mouse Monoclonal Antibody [clone: 5.8A]

Lieferant: Novus Biologicals
NOVUNB100-56511EA 761 CHF
NOVUNB100-56511
Anti-MYOD1 Mouse Monoclonal Antibody [clone: 5.8A]
Antikörper
Type: Primary
Antigen: MYOD1 (myogenic differentiation 1)
Clonality: monoclonal
Clone: 5.8A
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR