Anti-TERF2 Mouse Monoclonal Antibody [clone: 4A794.15]

Lieferant: Novus Biologicals
NOVUNB100-56506EA 803 CHF
NOVUNB100-56506
Anti-TERF2 Mouse Monoclonal Antibody [clone: 4A794.15]
Antikörper
Type: Primary
Antigen: TERF2 (telomeric repeat binding factor 2)
Clonality: monoclonal
Clone: 4A794.15
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR