Anti-IL27 Rabbit Polyclonal Antibody

Lieferant: Novus Biologicals
NOVUNB100-56389EA 495 CHF
NOVUNB100-56389
Anti-IL27 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: IL27 (interleukin 27)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR