Anti-RIPK1 Rabbit Polyclonal Antibody
Lieferant: Novus Biologicals
NOVUNB100-56160EA 412 CHF
NOVUNB100-56160
Anti-RIPK1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RIPK1 (receptor (TNFRSF)-interacting serine-threonine kinase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human