Anti-Cre Rabbit Polyclonal Antibody

Lieferant: Novus Biologicals
NOVUNB100-56135EA 581 CHF
NOVUNB100-56135
Anti-Cre Rabbit Polyclonal Antibody
Antikörper
The Cre Antibody from Novus Biologicals is a rabbit polyclonal antibody to Cre. This antibody reacts with cre/lox system. The Cre Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Recommended Dilutions: Immunohistochemistry (Parafin): 1/1000 - 1/5000

Type: Primary
Antigen: Cre
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Cre/lox System
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR