Anti-IgG Rabbit Polyclonal Antibody (CF™ 647)

Lieferant: Biotium

20285.EA 416 CHF
20285. 20285-1
Anti-IgG Rabbit Polyclonal Antibody (CF™ 647)
Antikörper
Type: Secondary
Antigen: IgG (Mouse) (mouse IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 647 Dye
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR