Anti-GFP Mouse Monoclonal Antibody (CF®488A)

Lieferant: Biotium

20215.EA 505 CHF
20215.
Anti-GFP Mouse Monoclonal Antibody (CF®488A)
Antikörper
Type: Tag
Antigen: GFP (green fluorescent protein)
Clonality: monoclonal
Clone:
Conjugation: CF® 488A Dye
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR