Anti-IgG Donkey Polyclonal Antibody (CF™ 488A)

Lieferant: Biotium

20166.EA 416 CHF
20166. 20166-1
Anti-IgG Donkey Polyclonal Antibody (CF™ 488A)
Antikörper
Type: Secondary
Antigen: IgG (Chicken) (chicken IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 488A Dye
Epitope:
Host: Donkey
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR