Anti-IgG Goat Polyclonal Antibody (CF™ 633)

Lieferant: Biotium

20129-1EA 140 CHF
20129-1 20129.
Anti-IgG Goat Polyclonal Antibody (CF™ 633)
Antikörper
Type: Secondary
Antigen: IgG (Guinea pig) (guinea pig IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 633 Dye
Epitope:
Host: Goat
Isotype: none
Reactivity: Guinea pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR