Anti-IgG Goat Polyclonal Antibody (CF™ 640R)

Lieferant: Biotium
20089 20089-1
20089.EA 360 CHF
20089. 20089-1
Anti-IgG Goat Polyclonal Antibody (CF™ 640R)
Antikörper
Type: Secondary
Antigen: IgG (Pig) (pig IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 640R Dye
Epitope:
Host: Goat
Isotype: none
Reactivity: Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR