Anti-IgG Rabbit Polyclonal Antibody (CF™ 680)

Lieferant: Biotium

20068-1EA 160 CHF
20068-1 20068.
Anti-IgG Rabbit Polyclonal Antibody (CF™ 680)
Antikörper
Type: Secondary
Antigen: IgG (Goat) (goat IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 680 Dye
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Goat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR