Anti-IgG Donkey Polyclonal Antibody (CF™ 680)

Lieferant: Biotium

20060-1EA 173 CHF
20060-1 20060.
Anti-IgG Donkey Polyclonal Antibody (CF™ 680)
Antikörper
Type: Secondary
Antigen: IgG (Goat) (goat IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 680 Dye
Epitope:
Host: Donkey
Isotype: none
Reactivity: Goat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR