Anti-TLS/FUS Rabbit Monoclonal Antibody [clone: EPR5812]

Lieferant: Abcam
AB124923-100 AB124923-1ML
ABCAAB124923-100EA 714 CHF
ABCAAB124923-100 ABCAAB124923-1ML
Anti-TLS/FUS Rabbit Monoclonal Antibody [clone: EPR5812]
Antikörper
Rabbit monoclonal [EPR5812] to TLS/FUS.

Bitte beachten All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: TLS/FUS
Clonality: Monoclonal
Clone: EPR5812
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR