Anti-ALP Mouse Monoclonal Antibody [clone: P4A10E4]

Lieferant: Abcam
ABCAAB116592-100EA 0 CHF
ABCAAB116592-100
Anti-ALP Mouse Monoclonal Antibody [clone: P4A10E4]
Antikörper
Type: Primary
Antigen: ALP (Alkaline Phosphatase)
Clonality: monoclonal
Clone: P4A10E4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR