Anti-Somatostatin 28 Rabbit Monoclonal Antibody [clone: EPR3360(2)]
Lieferant: Abcam
AB111050-100 AB111050-1ML
ABCAAB111050-100EA 630 CHF
ABCAAB111050-100 ABCAAB111050-1ML
Anti-Somatostatin 28 Rabbit Monoclonal Antibody [clone: EPR3360(2)]
Antikörper
Rabbit monoclonal [EPR3360(2)] to Somatostatin 28.
Bitte beachten All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Somatostatin 28
Clonality: Monoclonal
Clone: EPR3360(2)
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human