Anti-CDK2 Rabbit monoclonal antibody unconjugated [clone: SP80]

Lieferant: Abcam
AB101682-100 AB101682-500 AB101682-1
ABCAAB101682-100EA 660 CHF
ABCAAB101682-100 ABCAAB101682-500 ABCAAB101682-1
Anti-CDK2 Rabbit monoclonal antibody unconjugated [clone: SP80]
Antikörper
Bitte beachten

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.



Type: Primary
Antigen: CDK2 (cyclin-dependent kinase 2)
Clonality: monoclonal
Clone: SP80
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR