Anti-Lactate Dehydrogenase Rabbit Monoclonal Antibody [clone: EPR1564]

Lieferant: Abcam
AB101562-100 AB101562-1ML
ABCAAB101562-100EA 630 CHF
ABCAAB101562-100 ABCAAB101562-1ML
Anti-Lactate Dehydrogenase Rabbit Monoclonal Antibody [clone: EPR1564]
Antikörper
Rabbit monoclonal [EPR1564] to Lactate Dehydrogenase.

Bitte beachten All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Lactate Dehydrogenase
Clonality: Monoclonal
Clone: EPR1564
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR