Anti-IgG1 Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Abcam
ABCAAB97240-1EA 262.87 CHF
ABCAAB97240-1
Anti-IgG1 Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Type: Secondary
Antigen: IgG1 (Mouse) (mouse IgG1)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR