Anti-CS Rabbit polyclonal antibody unconjugated

Lieferant: Abcam
ABCAAB96600-100EA 582.07 CHF
ABCAAB96600-100
Anti-CS Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: CS (citrate synthase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR