Anti-IVD Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB95994-50EA 478.8 CHF
ABCAAB95994-50
Anti-IVD Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: IVD (isovaleryl-CoA dehydrogenase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR