Anti-Thymidine Kinase 1/TK1 Rabbit Monoclonal Antibody [clone: EPR3194]

Lieferant: Abcam
AB91651-100 AB91651-1ML
ABCAAB91651-100EA 690 CHF
ABCAAB91651-100 ABCAAB91651-1ML
Anti-Thymidine Kinase 1/TK1 Rabbit Monoclonal Antibody [clone: EPR3194]
Antikörper
Rabbit monoclonal [EPR3194] to Thymidine Kinase 1/TK1.

Bitte beachten All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Thymidine Kinase 1/TK1
Clonality: Monoclonal
Clone: EPR3194
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR