Anti-ZNF468 Mouse Polyclonal Antibody

Lieferant: Abcam
ABCAAB88862-50EA 478.8 CHF
ABCAAB88862-50
Anti-ZNF468 Mouse Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ZNF468 (zinc finger protein 468)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR