Anti-RNASEH2A Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB83943-100EA 525.74 CHF
ABCAAB83943-100
Anti-RNASEH2A Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RNASEH2A (ribonuclease H2, subunit A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR