Anti-Laminin 5 Mouse Monoclonal Antibody [clone: P3H9-2]

Lieferant: Abcam
ABCAAB78286-100EA 539.83 CHF
ABCAAB78286-100
Anti-Laminin 5 Mouse Monoclonal Antibody [clone: P3H9-2]
Antikörper
Type: Primary
Antigen: Laminin 5
Clonality: monoclonal
Clone: P3H9-2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR