Anti-STAT3 Rabbit Monoclonal Antibody [clone: EP2147Y]

Lieferant: Abcam
ABCAAB76315-100EA 619.63 CHF
ABCAAB76315-100
Anti-STAT3 Rabbit Monoclonal Antibody [clone: EP2147Y]
Antikörper
Type: Primary
Antigen: STAT3 (signal transducer and activator of transcription 3 (acute-phase response factor))
Clonality: monoclonal
Clone: EP2147Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR